4-(4-(Benzo[d][1,3]dioxol-5-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl)benzamide 99%

Supplier: Ambeed
Warning

A172016-1MG A172016-50MG A172016-250MG
76720-540EA 12 USD
76720-540 77550-110 77871-862
4-(4-(Benzo[d][1,3]dioxol-5-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl)benzamide 99%
4-(4-(Benzo[d][1,3]dioxol-5-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₄O₃
MDL Number: MFCD05865244
CAS Number: 301836-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR