Methyl stearate 99+%

Supplier: Ambeed
Warning

Synonyms: Stearic acid methyl ester, Methyl octadecanoate

A171330-100G A171330-25G A171330-500G
76720-534EA 14 USD
76720-534 76720-536 76720-538
Methyl stearate 99+%
Methyl stearate

Precaución: for Research Use Only

Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355
MDL Number: MFCD00009005
CAS Number: 112-61-8
Merck Index: 12,08959

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR