Sparfloxacin 99+%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-cyclopropyl-7-((3S,5R)-3,5-dimethylpiperazin-1-yl)-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

A170693-100G A170693-100MG A170693-1G A170693-5G A170693-1MG
76720-510EA 354 USD
76720-510 76720-512 76720-514 76720-516 77667-386
Sparfloxacin 99+%
Sparfloxacin

Precaución: for Research Use Only

Formula: C₁₉H₂₂F₂N₄O₃
MDL Number: MFCD00869619
CAS Number: 110871-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR