N-Methyl-DL-leucine 98%

Supplier: Ambeed
Warning

Synonyms: N-DL-Me-Leu-OH, N-Me-DL-Leu-OH

A170445-1G A170445-5G A170445-10G A170445-250MG
76720-502EA 67 USD
76720-502 76720-504 77879-138 77879-139
N-Methyl-DL-leucine 98%
N-Methyl-DL-leucine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MDL Number: MFCD00037758
CAS Number: 2566-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR