Dipotassium hexachloroosmate Os 38-40%

Supplier: Ambeed
Danger

Synonyms: Potassium hexachloroosmate(IV)

A170390-1G A170390-250MG
76720-498EA 77.45 USD
76720-498 76720-500
Dipotassium hexachloroosmate Os 38-40%
Dipotassium hexachloroosmate

Precaución: for Research Use Only

Formula: Cl₆K₂Os
MW: 481.14 g/mol
MDL Number: MFCD00011371
CAS Number: 16871-60-6
UN: 3290
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR