Phloretin 98%
Supplier: Ambeed
|
Warning
|
A170374-10G
A170374-1G
A170374-250MG
A170374-25G
A170374-50MG
A170374-5G
76720-484EA
33.95
USD
76720-484
76720-486
76720-488
76720-490
76720-492
77600-376
Phloretin 98%
Phloretin
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₅ MW: 274.28 g/mol |
MDL Number:
MFCD00002288 CAS Number: 60-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...