(3-Amino-1-hydroxypropane-1,1-diyl)bis(phosphonic acid) 98%

Supplier: Ambeed
Warning

Synonyms: Amidronate, Pamidronic acid

A169448-10MG A169448-1MG A169448-50MG
76720-434EA 12.74 USD
76720-434 76720-436 76720-438
(3-Amino-1-hydroxypropane-1,1-diyl)bis(phosphonic acid) 98%
(3-Amino-1-hydroxypropane-1,1-diyl)bis(phosphonic acid)

Precaución: for Research Use Only

Formula: C₃H₁₁NO₇P₂
MW: 235.07 g/mol
MDL Number: MFCD00168777
CAS Number: 40391-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR