Methyl-5-bromopyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

A168672-100MG A168672-1G A168672-250MG A168672-25G A168672-5G A168672-100G
76720-408EA 9 USD
76720-408 76720-410 76720-412 76720-414 77600-356 77879-133
Methyl-5-bromopyrazine-2-carboxylate 98%
Methyl 5-bromopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD09991715
CAS Number: 210037-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR