3-(1-Naphthyl)-D-alanine 97%

Supplier: Ambeed
Warning

Synonyms: H-D-1-Nal-OH

A168422-10G A168422-1G A168422-250MG A168422-25G A168422-5G
76720-386EA 83.82 USD
76720-386 76720-388 76720-390 76720-392 76720-394
3-(1-Naphthyl)-D-alanine 97%
3-(1-Naphthyl)-D-alanine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.252 g/mol
MDL Number: MFCD00038544
CAS Number: 78306-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR