Octyl 2-cyanoacetate 98%

Supplier: Ambeed
Warning

A168263-100G A168263-10G A168263-25G A168263-5G
76720-378EA 134.74 USD
76720-378 76720-380 76720-382 76720-384
Octyl 2-cyanoacetate 98%
Octyl 2-cyanoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MW: 197.27 g/mol
MDL Number: MFCD00075425
CAS Number: 15666-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR