Methyl-6-fluorosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-fluoro-6-hydroxybenzoate

A167055-100G A167055-100MG A167055-10G A167055-1G A167055-250MG A167055-25G A167055-5G
76720-338EA 315 USD
76720-338 76720-340 76720-342 76720-344 76720-346 76720-348 76720-350
Methyl-6-fluorosalicylate 98%
Methyl 6-fluorosalicylate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.1378 g/mol
MDL Number: MFCD00153207
CAS Number: 72373-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR