N-(Trimethylsilyl)acetamide 95%

Supplier: Ambeed
Danger

Synonyms: Acetic acid (trimethylsilyl)amide

A166960-25G A166960-5G
76720-334EA 44 USD
76720-334 76720-336
N-(Trimethylsilyl)acetamide 95%
N-(Trimethylsilyl)acetamide

Precaución: for Research Use Only

Formula: C₅H₁₃NOSi
MDL Number: MFCD00008671
CAS Number: 13435-12-6
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR