Mitoxantrone dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Novantrone, 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione dihydrochloride

A166750-10MG A166750-1MG A166750-50MG A166750-1G
76720-328EA 19 USD
76720-328 76720-330 76720-332 77879-116
Mitoxantrone dihydrochloride 98%
Mitoxantrone dihydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₃₀Cl₂N₄O₆
MW: 517.404 g/mol
MDL Number: MFCD00242943
CAS Number: 70476-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR