4-Morpholinyl(2-piperidinyl)methanone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Piperidin-2-ylcarbonyl)morpholine hydrochloride

A166361-1G A166361-5G
76720-324EA 15 USD
76720-324 76720-326
4-Morpholinyl(2-piperidinyl)methanone hydrochloride 97%
4-Morpholinyl(2-piperidinyl)methanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₉ClN₂O₂
MW: 234.73 g/mol
MDL Number: MFCD05865124
CAS Number: 690634-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR