Isobutyl myristate 98+%

Supplier: Ambeed
Warning

A165698-100G A165698-25G A165698-5G
76720-300EA 111 USD
76720-300 76720-302 76720-304
Isobutyl myristate 98+%
Isobutyl myristate

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₂
MDL Number: MFCD00059286
CAS Number: 25263-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR