O-Methylisourea hemisulfate 98,5%

Supplier: Ambeed
Warning

Synonyms: O-Methylisouronium sulphate

A164915-100G A164915-25G A164915-500G
76720-266EA 8.49 USD
76720-266 76720-268 76720-270
O-Methylisourea hemisulfate 98,5%
O-Methylisourea hemisulfate

Precaución: for Research Use Only

Formula: C₄H₁₄N₄O₆S
MW: 246.24 g/mol
MDL Number: MFCD00040594
CAS Number: 52328-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR