Disodium hexachloroplatinate hexahydrate 98%

Supplier: Ambeed
Danger

Synonyms: Sodium hexachloroplatinate(IV) hexahydrate, Sodium hexachloroplatinate (IV) hexahydrate

A164878-100MG A164878-1G
76720-262EA 19 USD
76720-262 76720-264
Disodium hexachloroplatinate hexahydrate 98%
Disodium hexachloroplatinate hexahydrate

Precaución: for Research Use Only

Formula: Cl₆Na₂Pt·6H₂O
MDL Number: MFCD00149172
CAS Number: 19583-77-8
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR