N-(Trimethylsilylmethyl)benzylamine 96%
Supplier: Ambeed
|
Warning
|
A163760-100G
A163760-10G
A163760-1G
A163760-25G
A163760-500G
A163760-5G
76720-240EA
84
USD
76720-240
76720-242
76720-244
76720-246
76720-248
76720-250
N-(Trimethylsilylmethyl)benzylamine 96%
N-(Trimethylsilylmethyl)benzylamine
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NSi |
MDL Number:
MFCD00012192 CAS Number: 53215-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...