Methyl-4-(bromomethyl)-3-chlorobenzoate 95%

Supplier: Ambeed
Danger

A163410-10G A163410-250MG A163410-5G A163410-1G
76720-220EA 286 USD
76720-220 76720-222 76720-224 77667-288
Methyl-4-(bromomethyl)-3-chlorobenzoate 95%
Methyl-4-(bromomethyl)-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MDL Number: MFCD10566565
CAS Number: 74733-30-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR