1-Bromo-4-cyclohexylbenzene 97%

Supplier: Ambeed
Warning

A163057-100G A163057-10G A163057-1G A163057-25G A163057-5G
76720-208EA 351.16 USD
76720-208 76720-210 76720-212 76720-214 76720-216
1-Bromo-4-cyclohexylbenzene 97%
1-Bromo-4-cyclohexylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₅Br
MW: 239.16 g/mol
MDL Number: MFCD00019339
CAS Number: 25109-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR