Repaglinide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic acid

A162841-100MG A162841-10G A162841-10MG A162841-1G A162841-250MG A162841-25G A162841-50MG A162841-5G
76720-186EA 12 USD
76720-186 76720-188 76720-190 76720-192 76720-194 76720-196 76720-198 76720-200
Repaglinide 98%
Repaglinide

Precaución: for Research Use Only

Formula: C₂₇H₃₆N₂O₄
MW: 452.59 g/mol
MDL Number: MFCD00906179
CAS Number: 135062-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR