Z-Lys(Z)-OH 97%

Supplier: Ambeed
Warning

A162555-100G A162555-25G A162555-500G A162555-5G
76720-160EA 96 USD
76720-160 76720-162 76720-164 76720-166 76720-168
Z-Lys(Z)-OH 97%
Z-Lys(Z)-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₆
MW: 414.458 g/mol
MDL Number: MFCD00037822
CAS Number: 405-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR