10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine 98%

Supplier: Ambeed
Warning

Synonyms: Quetiapine Related Compound G, Dibenzo[b,f][1,4]thiazepin-11(10H)-one

A161851-100G A161851-25G A161851-500G
76720-144EA 92.3 USD
76720-144 76720-146 76720-148
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine 98%
10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine

Precaución: for Research Use Only

Formula: C₁₃H₉NOS
MW: 227.2817 g/mol
MDL Number: MFCD00901197
CAS Number: 3159-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR