Ethyl-5,6-dimethoxyindole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5,6-dimethoxy-1H-indole-2-carboxylate

A160256-100MG A160256-1G A160256-250MG A160256-25G A160256-5G
76720-094EA 16.98 USD
76720-094 76720-096 76720-098 76720-100 76720-102
Ethyl-5,6-dimethoxyindole-2-carboxylate 95%
Ethyl-5,6-dimethoxyindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
MW: 249.2600 g/mol
MDL Number: MFCD00015457
CAS Number: 16382-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR