Benzo[h]quinoline 98%

Supplier: Ambeed
Warning

A160237-100G A160237-10G A160237-1G A160237-250MG A160237-25G A160237-5G
76720-082EA 265 USD
76720-082 76720-084 76720-086 76720-088 76720-090 76720-092
Benzo[h]quinoline 98%
Benzo[h]quinoline

Precaución: for Research Use Only

Formula: C₁₃H₉N
MDL Number: MFCD00004984
CAS Number: 230-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR