(R)-(-)-2-(Methoxymethyl)pyrrolidine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-2-(Methoxymethyl)pyrrolidine
A159662-1G
A159662-250MG
76720-070EA
46.68
USD
76720-070
76720-072
(R)-(-)-2-(Methoxymethyl)pyrrolidine 98%
(R)-(-)-2-(Methoxymethyl)pyrrolidine
Precaución: for Research Use Only
Formula:
C₆H₁₃NO MW: 115.18 g/mol |
MDL Number:
MFCD00066219 CAS Number: 84025-81-0 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...