Methyl-3-hydroxy-4-methoxybenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-hydroxy-4-methoxybenzoate, Methyl isovanillate

A159335-100G A159335-10G A159335-1G A159335-25G A159335-5G A159335-500G
76720-054EA 130 USD
76720-054 76720-056 76720-058 76720-060 76720-062 77667-244
Methyl-3-hydroxy-4-methoxybenzoate 97%
Methyl-3-hydroxy-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
MDL Number: MFCD01321262
CAS Number: 6702-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR