4-Chloro-6,7-difluoro-2-methylquinazoline 95%

Supplier: Ambeed
Warning

A159240-100MG A159240-1G A159240-250MG
76720-044EA 55.17 USD
76720-044 76720-046 76720-048
4-Chloro-6,7-difluoro-2-methylquinazoline 95%
4-Chloro-6,7-difluoro-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₅ClF₂N₂
MW: 214.59 g/mol
MDL Number: MFCD06657231
CAS Number: 887592-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR