Pemetrexed 99+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo-[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, Alimta, [3H]-, N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid

A159122-100MG A159122-1G A159122-1MG A159122-250MG A159122-50MG A159122-5G
76720-034EA 74.27 USD
76720-034 76720-036 76720-038 76720-040 76720-042 77592-936
Pemetrexed 99+%
Pemetrexed

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₅O₆
MW: 427.42 g/mol
MDL Number: MFCD19105065
CAS Number: 137281-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR