2-Benzyloxy-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Benzyloxy)-4-fluorophenyl)boronic acid, 2-Benzyloxy-4-fluorobenzeneboronic acid

A158783-1G A158783-250MG A158783-5G
76720-028EA 20 USD
76720-028 76720-030 76720-032
2-Benzyloxy-4-fluorophenylboronic acid 98%
2-Benzyloxy-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD03788402
CAS Number: 848779-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR