Methyl-3-fluoro-4-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-fluoro-4-nitrobenzoate, Methyl 3-fluoro-4-nitrobenzenecarboxylate, 3-Fluorobenzoic acid methyl ester

A158192-100G A158192-10G A158192-1G A158192-250MG A158192-25G A158192-5G
76720-006EA 209 USD
76720-006 76720-008 76720-010 76720-012 76720-014 76720-016
Methyl-3-fluoro-4-nitrobenzoate 98%
Methyl-3-fluoro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
MDL Number: MFCD08444027
CAS Number: 185629-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR