Polydatin 99+%

Supplier: Ambeed
Warning

Synonyms: (E/Z)-Piceid, Reservatrol 3-β-mono-D-Glucoside, (E/Z)-Polydatin, Resveratrol 3-O-β-D-Glucopyranoside, Piceid, (2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-(4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, 3,4′-5-Trihydroxystilbene-3-β-D-Glucopyranoside, Trihydroxystilbene-3-β-D-Glucopyranoside

A157731-100G A157731-250MG A157731-25G A157731-500G A157731-5G A157731-1G A157731-50MG
76719-986EA 166.57 USD
76719-986 76719-988 76719-990 76719-992 76719-994 77600-246 77667-222
Polydatin 99+%
Polydatin

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₈
MW: 390.3839 g/mol
MDL Number: MFCD00210592
CAS Number: 65914-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR