Pimobendan 98%

Supplier: Ambeed
Warning

A157577-10MG A157577-250MG A157577-50MG A157577-1G A157577-5G
76719-976EA 5 USD
76719-976 76719-978 76719-980 77879-083 77879-084
Pimobendan 98%
Pimobendan

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₄O₂
MDL Number: MFCD00761648
CAS Number: 74150-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR