N-(4-(4-(4-Fluorophenyl)piperazin-1-yl)butan-2-yl)nicotinamide 95%

Supplier: Ambeed
Warning

A156843-1G A156843-250MG A156843-5G
76719-962EA 92.3 USD
76719-962 76719-964 76719-966
N-(4-(4-(4-Fluorophenyl)piperazin-1-yl)butan-2-yl)nicotinamide 95%
N-(4-(4-(4-Fluorophenyl)piperazin-1-yl)butan-2-yl)nicotinamide

Precaución: for Research Use Only

Formula: C₂₀H₂₅FN₄O
MW: 356.43 g/mol
MDL Number: MFCD00867991
CAS Number: 27367-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR