Phytic acid 60% in water

Supplier: Ambeed
Warning

Synonyms: Inositol hexaphosphoric acid

A155640-1000G A155640-100G A155640-25G A155640-500G
76719-926EA 42.44 USD
76719-926 76719-928 76719-930 76719-932
Phytic acid 60% in water
Phytic acid

Precaución: for Research Use Only

Formula: C₆H₁₈O₂₄P₆
MW: 660.03 g/mol
MDL Number: MFCD00082309
CAS Number: 83-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR