5-Chloro-6-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A155606-250MG A155606-100MG
76719-924EA 31 USD
76719-924 77879-075
5-Chloro-6-methylpyrimidin-4-amine 95%
5-Chloro-6-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MDL Number: MFCD09837292
CAS Number: 13040-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR