Sodium-3-(1-naphthylamino)propanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: N-(1-Naphthyl)-3-aminopropanesulphonic acid Sodium Salt

A154915-1G A154915-250MG A154915-25G A154915-5G
76719-892EA 19.1 USD
76719-892 76719-894 76719-896 76719-898
Sodium-3-(1-naphthylamino)propanesulfonate 98%
Sodium-3-(1-naphthylamino)propanesulphonate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃S
MW: 287.31 g/mol
MDL Number: MFCD00059520
CAS Number: 104484-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR