Methyl 2-hexenoate 97%

Supplier: Ambeed
Danger

A154352-10G A154352-5G A154352-25G
76719-866EA 22 USD
76719-866 76719-868 77879-062
Methyl 2-hexenoate 97%
Methyl 2-hexenoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MDL Number: MFCD00048798
CAS Number: 2396-77-2
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR