N-Carbobenzyloxy-D-proline 97%

Supplier: Ambeed
Warning

A154146-10G A154146-25G A154146-500G A154146-100G
76719-848EA 5 USD
76719-848 76719-850 76719-852 77567-242
N-Carbobenzyloxy-D-proline 97%
N-Carbobenzyloxy-D-proline

Precaución: For research use only.

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD00063228
CAS Number: 6404-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR