ɑ-(Trichloromethyl)benzyl acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2,2-Trichloro-1-phenylethyl acetate, Acetic acid 2,2,2-Trichloro-1-phenylethyl Ester, Acetic Acid ɑ-(Trichloromethyl)benzyl Ester, Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester, 2,2,2-Trichloro-1-phenylethyl Acetate, Acetic acid α-(Trichloromethyl)benzyl Ester, α-(Trichloromethyl)benzyl Acetate
A153958-100G
A153958-25G
A153958-500G
76719-842EA
15.92
USD
76719-842
76719-844
76719-846
ɑ-(Trichloromethyl)benzyl acetate 98%
alpha-(Trichloromethyl)benzyl acetate
Precaución: for Research Use Only
Formula:
C₁₀H₉Cl₃O₂ MW: 267.53 g/mol |
MDL Number:
MFCD00000804 CAS Number: 90-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...