4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Trifluoromethoxyphenoxy)piperidine
A153864-100G
A153864-10G
A153864-1G
A153864-25G
A153864-5G
76719-826EA
770.22
USD
76719-826
76719-828
76719-830
76719-832
76719-834
4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%
4-[4-(Trifluoromethoxy)phenoxy]piperidine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄F₃NO₂ MW: 261.25 g/mol |
MDL Number:
MFCD06656166 CAS Number: 287952-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...