Ethyl-2-(triphenylphosphoranylidene)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

A153553-100G A153553-10G A153553-5G A153553-25G
76719-814EA 213.25 USD
76719-814 76719-816 76719-818 76719-820 76719-822 77318-368
Ethyl-2-(triphenylphosphoranylidene)acetate 97%
Ethyl-2-(triphenylphosphoranylidene)acetate

Precaución: for Research Use Only

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
MDL Number: MFCD00009183
CAS Number: 1099-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR