2-(2-Pyridyl)indole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yl)-1H-indole

A152749-100MG A152749-1G A152749-250MG
76719-802EA 43.5 USD
76719-802 76719-804 76719-806
2-(2-Pyridyl)indole 98%
2-(2-Pyridyl)indole

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
MDL Number: MFCD00033466
CAS Number: 13228-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR