Prulifloxacin 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1-methyl-7-(4-((5-methyl-2-oxo-1,3-dioxol-4-yl)methyl)piperazin-1-yl)-4-oxo-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid

A151816-100MG A151816-1G A151816-250MG A151816-25MG
76719-768EA 14.86 USD
76719-768 76719-770 76719-772 76719-774
Prulifloxacin 98%
Prulifloxacin

Precaución: for Research Use Only

Formula: C₂₁H₂₀FN₃O₆S
MW: 461.46 g/mol
MDL Number: MFCD00864847
CAS Number: 123447-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR