Sodium stearoyl lactylate 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 2-((2-(stearoyloxy)propanoyl)oxy)propanoate

A151550-1000G A151550-10G A151550-1G A151550-25G A151550-500G A151550-5G A151550-100G
76719-750EA 125.19 USD
76719-750 76719-752 76719-754 76719-756 76719-758 76719-760 77592-846
Sodium stearoyl lactylate 97%
Sodium stearoyl lactylate

Precaución: for Research Use Only

Formula: C₂₄H₄₄O₆Na
MW: 450.5844 g/mol
MDL Number: MFCD00152806
CAS Number: 25383-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR