Ruthenium(III) chloride hydrate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate
A151048-10G
A151048-1G
A151048-5G
76719-724EA
242.95
USD
76719-724
76719-726
76719-728
Ruthenium(III) chloride hydrate 98%
Ruthenium(III) chloride hydrate
Precaución: for Research Use Only
Formula:
Cl₃Ru·xH₂O MW: 207.43 g/mol (anhy) |
MDL Number:
MFCD00149844 CAS Number: 14898-67-0 UN: 3260 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...