Methyl-3-ethoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-ethoxybenzoate

A151004-1G A151004-25G A151004-5G
76719-718EA 13.8 USD
76719-718 76719-720 76719-722
Methyl-3-ethoxybenzoate 98%
Methyl 3-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2000 g/mol
MDL Number: MFCD00017263
CAS Number: 108593-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR