Methyl-3-bromo-5-fluorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-5-fluorobenzoate

A150379-100G A150379-10G A150379-1G A150379-250MG A150379-25G A150379-5G
76719-690EA 162 USD
76719-690 76719-692 76719-694 76719-696 76719-698 76719-700
Methyl-3-bromo-5-fluorobenzoate 97%
Methyl-3-bromo-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD07780735
CAS Number: 334792-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR