4-Amino-4'-methyldiphenyl ether 98%

Supplier: Ambeed
Warning

Synonyms: 4-(p-Tolyloxy)aniline, 4-Aminophenyl p-Tolyl Ether 4-(p-Tolyloxy)aniline, 4-Amino-4'-methyldiphenyl Ether, 4-Aminophenyl p-Tolyl Ether

A149498-1G A149498-25G A149498-5G
76719-680EA 17 USD
76719-680 76719-682 76719-684
4-Amino-4'-methyldiphenyl ether 98%
4-Amino-4'-methyldiphenyl ether

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MDL Number: MFCD00059858
CAS Number: 41295-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR