4,4',4''-Trimethoxytrityl chloride 95%

Supplier: Ambeed
Warning

Synonyms: 4,4',4''-(Chloromethanetriyl)tris(methoxybenzene)

A148863-250MG A148863-1G A148863-5G
76719-670EA 14.86 USD
76719-670 77567-100 77667-122
4,4',4''-Trimethoxytrityl chloride 95%
4,4',4''-Trimethoxytrityl chloride

Precaución: For research use only.

Formula: C₂₂H₂₁ClO₃
MW: 368.8534 g/mol
MDL Number: MFCD00008408
CAS Number: 49757-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR